siRNA Details
| Gene Symbol: SLC45A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033102.2 | NP_149093.1 | 3 | 1213 | ||
| XM_005245556.3 | XP_005245613.1 | 3 | 1379 | |||
| XM_005245557.4 | XP_005245614.1 | 3 | 1305 | |||
| XM_005245560.2 | XP_005245617.1 | 3 | 1213 | |||
| XM_017002638.1 | XP_016858127.1 | 3 | 1752 | |||
| GenBank mRNA | AK290364.1 | - | 3 | 1214 | ||
| AK313479.1 | - | 3 | 1204 | |||
| AY033593.1 | - | 3 | 1157 | |||
| BC050416.2 | - | 3 | 1121 | 6198823 | ||

SLC45A3
Genomic Map