siRNA Details
| Gene Symbol: SEC16B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033127.2 | NP_149118.2 | 3 | 445 | ||
| GenBank mRNA | AB063357.1 | - | 3 | 445 | ||
| AK314585.1 | - | 3 | 511 | |||
| BC009106.1 | - | 3 | 437 | 3448742 | ||
| EF125213.1 | - | 2 | 355 | |||

SEC16B
Genomic Map