siRNA Details
| Gene Symbol: ESCO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052911.2 | NP_443143.2 | 4 | 1162 | ||
| XM_011525798.1 | XP_011524100.1 | 4 | 1292 | |||
| XM_011525799.2 | XP_011524101.1 | 4 | 1425 | |||
| GenBank mRNA | AB067498.1 | - | 4 | 1098 | ||
| AK096746.1 | - | 4 | 1164 | |||
| AL832041.1 | - | 4 | 1314 | |||
| BC089426.1 | - | 1 | 582 | 3090307 | ||

ESCO1
Genomic Map