siRNA Details
| Gene Symbol: TMEM123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052932.2 | NP_443164.2 | 5 | 1428 | ||
| GenBank mRNA | AK075420.1 | - | 5 | 1211 | ||
| AK225790.1 | - | 5 | 1398 | |||
| AL110202.1 | - | 1 | 367 | |||
| AY008283.1 | - | 4 | 1165 | |||
| AY358696.1 | - | 5 | 1098 | |||
| BC032296.1 | - | 5 | 1114 | 4340763 | ||

TMEM123
Genomic Map