siRNA Details
| Gene Symbol: LRRC42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256409.1 | NP_001243338.1 | 3, 4 | 753 | ||
| NM_052940.4 | NP_443172.1 | 2, 3 | 719 | |||
| XM_006710327.3 | XP_006710390.1 | 3, 4 | 575 | |||
| XM_006710328.3 | XP_006710391.1 | 3, 4 | 1088 | |||
| GenBank mRNA | AK074521.1 | - | 2, 3 | 692 | ||
| AK075201.1 | - | 2, 3 | 681 | |||
| AK090881.1 | - | 3, 4 | 1002 | |||
| AK095767.1 | - | 3, 4 | 672 | |||
| BC013101.1 | - | 2, 3 | 558 | 3918785 | ||
| CX783955.1 | - | 3, 4 | 753 | |||

LRRC42
Genomic Map