siRNA Details
Gene Symbol: MRGPRX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054031.3 | NP_473372.3 | 3 | 1392 | ||
XM_011519882.2 | XP_011518184.1 | 2 | 1247 | |||
GenBank mRNA | BC067292.1 | - | 3 | 1392 | 30383708 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
