siRNA Details
| Gene Symbol: GLYATL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001220494.2 | NP_001207423.1 | 3 | 332 | ||
| NM_001220496.2 | NP_001207425.1 | 2 | 160 | |||
| NM_080661.4 | NP_542392.2 | 2 | 160 | |||
| XM_011545354.2 | XP_011543656.1 | 2 | 276 | |||
| XM_011545355.2 | XP_011543657.1 | 2 | 262 | |||
| XM_011545356.2 | XP_011543658.1 | 3 | 321 | |||
| XM_011545357.2 | XP_011543659.1 | 3 | 307 | |||
| XM_011545358.2 | XP_011543660.1 | 3 | 845 | |||
| XM_011545359.2 | XP_011543661.1 | 2 | 801 | |||
| GenBank mRNA | AF087878.1 | - | 2 | 195 | ||
| AK055818.1 | - | 1 | 317 | |||
| AK091965.1 | - | 2 | 160 | |||
| AK300479.1 | - | 3 | 297 | |||
| BC008353.1 | - | 3 | 260 | 3537224 | ||
| DC339353.1 | - | 2 | 287 | |||
| DQ084381.1 | - | 2 | 160 | |||

GLYATL1
Genomic Map