siRNA Details
| Gene Symbol: KIAA2013 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138346.2 | NP_612355.1 | 3 | 2272 | ||
| GenBank mRNA | AL833891.1 | - | 1 | 1038 | ||
| BC004501.2 | - | 2 | 885 | |||
| BC035033.1 | - | 3 | 2275 | 5264442 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIAA2013| Gene Symbol: KIAA2013 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138346.2 | NP_612355.1 | 3 | 2272 | ||
| GenBank mRNA | AL833891.1 | - | 1 | 1038 | ||
| BC004501.2 | - | 2 | 885 | |||
| BC035033.1 | - | 3 | 2275 | 5264442 | ||