siRNA Details
| Gene Symbol: PIH1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082619.1 | NP_001076088.1 | 2 | 384 | ||
| NM_138789.3 | NP_620144.1 | 2 | 384 | |||
| XM_011542593.2 | XP_011540895.2 | 2 | 290 | |||
| XM_017017201.1 | XP_016872690.1 | 2 | 501 | |||
| XM_017017202.1 | XP_016872691.1 | 2 | 501 | |||
| XM_017017203.1 | XP_016872692.1 | 2 | 501 | |||
| XM_017017204.1 | XP_016872693.1 | 2 | 501 | |||
| XM_017017205.1 | XP_016872694.1 | 2 | 501 | |||
| GenBank mRNA | AK300496.1 | - | 2 | 247 | ||
| BC019238.2 | - | 1 | 193 | 3935322 | ||
| CB159963.1 | - | 2 | 384 | |||

PIH1D2
Genomic Map