siRNA Details
| Gene Symbol: DEPDC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077242.1 | NP_001070710.1 | 2 | 263 | ||
| NM_139160.2 | NP_631899.2 | 2 | 502 | |||
| GenBank mRNA | AF447588.1 | - | 2 | 502 | ||
| AJ245600.1 | - | 2 | 218 | |||
| AK294934.1 | - | 2 | 251 | |||
| AK295960.1 | - | 2 | 216 | |||
| BC030970.1 | - | 2 | 263 | 4133262 | ||

DEPDC7
Genomic Map