siRNA Details
| Gene Symbol: C1orf105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300760.1 | NP_001287689.1 | 5 | 633 | ||
| NM_139240.3 | NP_640333.3 | 7 | 732 | |||
| XM_005245608.2 | XP_005245665.1 | 7 | 596 | |||
| XM_005245610.3 | XP_005245667.1 | 6 | 513 | |||
| XM_006711630.2 | XP_006711693.1 | 5 | 419 | |||
| XM_011510152.2 | XP_011508454.1 | 9 | 2941 | |||
| XM_011510153.2 | XP_011508455.1 | 8 | 2904 | |||
| XM_011510154.2 | XP_011508456.1 | 6 | 2774 | |||
| XM_011510155.2 | XP_011508457.1 | 8 | 2873 | |||
| XM_011510156.2 | XP_011508458.1 | 5 | 2683 | |||
| XM_017002808.1 | XP_016858297.1 | 7 | 530 | |||
| GenBank mRNA | AL035295.1 | - | 7 | 732 | ||
| BC038410.1 | - | 7 | 730 | 5169137 | ||

C1orf105
Genomic Map