siRNA Details
| Gene Symbol: ANGEL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300753.1 | NP_001287682.1 | 3 | 533 | ||
| NM_001300755.1 | NP_001287684.1 | 3 | 631 | |||
| NM_001300757.1 | NP_001287686.1 | 2 | 286 | |||
| NM_001300758.1 | NP_001287687.1 | 2 | 282 | |||
| NM_144567.4 | NP_653168.2 | 3 | 608 | |||
| NR_125333.1 | 2 | 282 | ||||
| XM_005273344.1 | XP_005273401.1 | 3 | 629 | |||
| XM_005273345.1 | XP_005273402.1 | 3 | 675 | |||
| XM_005273346.2 | XP_005273403.1 | 3 | 428 | |||
| XM_005273347.3 | XP_005273404.1 | 2 | 300 | |||
| XM_017002774.1 | XP_016858263.1 | 3 | 557 | |||
| XM_017002775.1 | XP_016858264.1 | 4 | 856 | |||
| XM_017002776.1 | XP_016858265.1 | 4 | 880 | |||
| XM_017002777.1 | XP_016858266.1 | 4 | 926 | |||
| XM_017002778.1 | XP_016858267.1 | 4 | 669 | |||
| GenBank mRNA | AF510741.1 | - | 2 | 237 | ||
| AK294479.1 | - | 3 | 407 | |||
| AK295116.1 | - | 3 | 537 | |||
| AK299208.1 | - | 3 | 600 | |||
| AK299365.1 | - | 2 | 192 | |||
| AK311276.1 | - | 2 | 191 | |||
| AL079275.1 | - | 3 | 563 | |||
| AY468376.1 | - | 2 | 338 | |||
| BC016966.1 | - | 2 | 352 | |||
| BC047469.1 | - | 2 | 198 | 5274835 | ||

ANGEL2
Genomic Map