siRNA Details
| Gene Symbol: DYRK1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004714.2 | NP_004705.1 | 2, 3 | 383 | ||
| NM_006483.2 | NP_006474.1 | 2, 3 | 383 | |||
| NM_006484.2 | NP_006475.1 | 2, 3 | 383 | |||
| XM_005259398.4 | XP_005259455.1 | 2, 3 | 835 | |||
| GenBank mRNA | AF205861.1 | - | 2, 3 | 280 | ||
| AK074971.1 | - | 2, 3 | 299 | |||
| AK302127.1 | - | 2, 3 | 312 | |||
| BC018751.1 | - | 2, 3 | 335 | 4300012 | ||
| BC025291.1 | - | 2, 3 | 286 | 5019417 | ||
| HY024135.1 | - | 2, 3 | 384 | |||
| Y17999.1 | - | 2, 3 | 351 | |||

DYRK1B
Genomic Map