siRNA Details
| Gene Symbol: BROX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288579.1 | NP_001275508.1 | 6 | 850 | ||
| NM_001288580.1 | NP_001275509.1 | 6 | 556 | |||
| NM_001288581.1 | NP_001275510.1 | 7 | 652 | |||
| NM_144695.3 | NP_653296.2 | 7 | 946 | |||
| XM_005273065.2 | XP_005273122.2 | 7 | 582 | |||
| XM_005273069.4 | XP_005273126.1 | 6 | 818 | |||
| XM_006711173.3 | XP_006711236.1 | 8 | 1015 | |||
| XM_011509212.2 | XP_011507514.1 | 6 | 909 | |||
| XM_011509213.2 | XP_011507515.1 | 6 | 890 | |||
| XM_011509214.2 | XP_011507516.1 | 8 | 815 | |||
| XM_017000374.1 | XP_016855863.1 | 9 | 1140 | |||
| XM_017000375.1 | XP_016855864.1 | 5 | 795 | |||
| XM_017000376.1 | XP_016855865.1 | 7 | 943 | |||
| GenBank mRNA | AB276123.1 | - | 6 | 540 | ||
| AK056983.1 | - | 7 | 935 | |||
| AK293974.1 | - | 6 | 482 | |||
| AK315930.1 | - | 6 | 486 | |||
| BC113635.1 | - | 7 | 683 | 8322687 | ||
| BC113637.1 | - | 7 | 683 | 8322689 | ||
| BC143811.1 | - | 6 | 587 | 9052326 | ||

BROX
Genomic Map