siRNA Details
| Gene Symbol: CIART | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300838.1 | NP_001287767.1 | 2 | 383 | ||
| NM_001300839.1 | NP_001287768.1 | 2 | 129 | |||
| NM_001300841.1 | NP_001287770.1 | 2 | 145 | |||
| NM_144697.3 | NP_653298.1 | 1 | 972 | |||
| XM_017000378.1 | XP_016855867.1 | 2 | 314 | |||
| GenBank mRNA | AK056960.1 | - | 1 | 253 | ||
| AK098755.1 | - | 1 | 656 | |||
| AK315397.1 | - | 1 | 657 | |||
| BC017397.1 | - | 2 | 101 | |||
| BC027999.1 | - | 2 | 373 | 5194413 | ||
| BC047238.1 | - | 1 | 686 | 5273646 | ||
| BE784098.1 | - | 2 | 75 | |||

CIART
Genomic Map