siRNA Details
| Gene Symbol: ALG14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001305242.1 | NP_001292171.1 | 5 | 619 | ||
| NM_144988.3 | NP_659425.1 | 4 | 582 | |||
| NR_131032.1 | 3 | 430 | ||||
| GenBank mRNA | AK289395.1 | - | 4 | 582 | ||
| BC011706.2 | - | 4 | 558 | 3689162 | ||
| HY073066.1 | - | 3 | 394 | |||

ALG14
Genomic Map