siRNA Details
| Gene Symbol: SLFNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168247.2 | NP_001161719.1 | 3 | 776 | ||
| NM_001300859.1 | NP_001287788.1 | 3 | 683 | |||
| NM_144990.3 | NP_659427.3 | 3 | 620 | |||
| XM_005270597.2 | XP_005270654.1 | 2 | 1359 | |||
| XM_005270598.2 | XP_005270655.1 | 2 | 675 | |||
| XM_005270599.2 | XP_005270656.1 | 3 | 683 | |||
| XM_006710432.3 | XP_006710495.1 | 2 | 1770 | |||
| XM_011540943.1 | XP_011539245.1 | 2 | 1306 | |||
| XM_011540945.1 | XP_011539247.1 | 2 | 1243 | |||
| XM_011540947.1 | XP_011539249.1 | 2 | 768 | |||
| XM_011540948.1 | XP_011539250.1 | 3 | 1221 | |||
| XM_011540949.1 | XP_011539251.1 | 2 | 1151 | |||
| XM_011540950.1 | XP_011539252.1 | 2 | 1121 | |||
| XM_011540951.1 | XP_011539253.1 | 3 | 675 | |||
| XM_011540952.2 | XP_011539254.1 | 2 | 1098 | |||
| XM_011540953.2 | XP_011539255.1 | 2 | 1678 | |||
| XM_011540954.1 | XP_011539256.1 | 2 | 1029 | |||
| XM_017000573.1 | XP_016856062.1 | 2 | 1359 | |||
| XM_017000574.1 | XP_016856063.1 | 2 | 1359 | |||
| GenBank mRNA | AK097609.1 | - | 3 | 671 | ||
| AK292296.1 | - | 3 | 746 | |||
| BC022037.2 | - | 3 | 582 | 4826722 | ||
| BC037879.1 | - | 1 | 1432 | 5274032 | ||
| BC050353.1 | - | 1 | 2940 | 5265392 | ||

SLFNL1
Genomic Map