siRNA Details
| Gene Symbol: MYOM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152372.3 | NP_689585.3 | 19 | 2470 | ||
| GenBank mRNA | AK128233.1 | - | 11 | 1530 | ||
| BC067101.1 | - | 19 | 2480 | 5928128 | ||
| BX537911.1 | - | 18 | 2194 | |||
| BX647453.1 | - | 19 | 2431 | |||
| BX647455.1 | - | 19 | 2470 | |||
| BX647644.1 | - | 19 | 2431 | |||

MYOM3
Genomic Map