siRNA Details
| Gene Symbol: MYOM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152372.3 | NP_689585.3 | 25 | 3271 | ||
| GenBank mRNA | AK128233.1 | - | 17 | 2331 | ||
| BC067101.1 | - | 25 | 3281 | 5928128 | ||
| BX537911.1 | - | 24 | 2995 | |||
| BX647453.1 | - | 25 | 3232 | |||
| BX647455.1 | - | 25 | 3271 | |||
| BX647644.1 | - | 25 | 3232 | |||

MYOM3
Genomic Map