siRNA Details
| Gene Symbol: C1orf131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300830.1 | NP_001287759.1 | 3, 4 | 562 | ||
| NM_152379.3 | NP_689592.2 | 3, 4 | 565 | |||
| GenBank mRNA | AK303359.1 | - | 3, 4 | 521 | ||
| AL834274.1 | - | 3, 4 | 548 | |||
| BC036800.1 | - | 3, 4 | 542 | 5589128 | ||
| BC053609.1 | - | 3, 4 | 514 | |||
| BC062353.1 | - | 3, 4 | 545 | 4648754 | ||

C1orf131
Genomic Map