siRNA Details
| Gene Symbol: B3GALNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277155.2 | NP_001264084.1 | 3 | 549 | ||
| NM_152490.4 | NP_689703.1 | 2 | 426 | |||
| XM_006711749.3 | XP_006711812.1 | 2 | 368 | |||
| XM_017000394.1 | XP_016855883.1 | 3 | 549 | |||
| XM_017000395.1 | XP_016855884.1 | 3 | 502 | |||
| GenBank mRNA | AK091497.1 | - | 2 | 199 | ||
| AK130297.1 | - | 2 | 370 | |||
| BC016974.1 | - | 3 | 490 | |||
| BC029564.1 | - | 2 | 426 | 4826983 | ||

B3GALNT2
Genomic Map