siRNA Details
| Gene Symbol: FAM89B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098784.1 | NP_001092254.1 | 2 | 1267 | ||
| NM_001098785.1 | NP_001092255.1 | 2 | 1257 | |||
| NM_152832.2 | NP_690045.1 | 2 | 1218 | |||
| GenBank mRNA | AF052151.1 | - | 2 | 1154 | ||
| BC023991.2 | - | 3 | 1665 | 2819240 | ||
| BC032373.1 | - | 2 | 1229 | 5123262 | ||
| BQ927525.1 | - | 1 | 458 | |||

FAM89B
Genomic Map