siRNA Details
| Gene Symbol: TMEM86A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153347.2 | NP_699178.1 | 3 | 2722 | ||
| GenBank mRNA | BC035692.1 | - | 3 | 2653 | 5759114 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM86A| Gene Symbol: TMEM86A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153347.2 | NP_699178.1 | 3 | 2722 | ||
| GenBank mRNA | BC035692.1 | - | 3 | 2653 | 5759114 | |