siRNA Details
| Gene Symbol: ZCCHC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153367.3 | NP_699198.2 | 4 | 4511 | ||
| GenBank mRNA | AL049949.1 | - | 1 | 2321 | ||
| BC028617.1 | - | 4 | 4513 | 4821347 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZCCHC24| Gene Symbol: ZCCHC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153367.3 | NP_699198.2 | 4 | 4511 | ||
| GenBank mRNA | AL049949.1 | - | 1 | 2321 | ||
| BC028617.1 | - | 4 | 4513 | 4821347 | ||