siRNA Details
| Gene Symbol: TEDDM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172000.3 | NP_741997.3 | 1 | 310 | ||
| GenBank mRNA | AJ515384.2 | - | 1 | 310 | ||
| GQ472227.1 | - | 1 | 310 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TEDDM1| Gene Symbol: TEDDM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172000.3 | NP_741997.3 | 1 | 310 | ||
| GenBank mRNA | AJ515384.2 | - | 1 | 310 | ||
| GQ472227.1 | - | 1 | 310 | |||