siRNA Details
Gene Symbol: TEDDM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172000.3 | NP_741997.3 | 1 | 854 | ||
GenBank mRNA | AJ515384.2 | - | 1 | 854 | ||
GQ472227.1 | - | 1 | 854 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TEDDM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172000.3 | NP_741997.3 | 1 | 854 | ||
GenBank mRNA | AJ515384.2 | - | 1 | 854 | ||
GQ472227.1 | - | 1 | 854 |