siRNA Details
| Gene Symbol: C1orf127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170754.1 | NP_001164225.1 | 9 | 902 | ||
| XM_017000373.1 | XP_016855862.1 | 7 | 754 | |||
| GenBank mRNA | AK094437.1 | - | 4 | 406 | ||
| BC126349.1 | - | 5 | 492 | 8992065 | ||
| BC143794.1 | - | 5 | 492 | 9052308 | ||
| BC143795.1 | - | 5 | 492 | 9052309 | ||
| BC143797.1 | - | 5 | 492 | 9052311 | ||

C1orf127
Genomic Map