siRNA Details
| Gene Symbol: C11orf44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271983.1 | NP_001258912.1 | 3 | 1190 | ||
| GenBank mRNA | AK096377.1 | - | 3 | 1190 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C11orf44| Gene Symbol: C11orf44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271983.1 | NP_001258912.1 | 3 | 1190 | ||
| GenBank mRNA | AK096377.1 | - | 3 | 1190 | ||