siRNA Details
| Gene Symbol: KLHL35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039548.2 | NP_001034637.2 | 3 | 1165 | ||
| XM_011544954.2 | XP_011543256.1 | 4 | 2674 | |||
| GenBank mRNA | AK091109.1 | - | 3 | 622 | ||
| BC042952.2 | - | 3 | 706 | 5275230 | ||
| BC057763.1 | - | 3 | 727 | |||
| BC132708.1 | - | 3 | 529 | 40146730 | ||
| BC132710.1 | - | 3 | 529 | 40146732 | ||

KLHL35
Genomic Map