siRNA Details
| Gene Symbol: RNF207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK056658.1 | - | 8 | 914 | ||
| AK294223.1 | - | 10 | 1018 | |||
| BC128237.1 | - | 9 | 912 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNF207| Gene Symbol: RNF207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK056658.1 | - | 8 | 914 | ||
| AK294223.1 | - | 10 | 1018 | |||
| BC128237.1 | - | 9 | 912 | |||