siRNA Details
| Gene Symbol: CCL21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002989.3 | NP_002980.1 | 2 | 196 | ||
| XM_011518004.2 | XP_011516306.1 | 2 | 183 | |||
| GenBank mRNA | AB002409.1 | - | 2 | 153 | ||
| AF001979.1 | - | 2 | 95 | |||
| AK311901.1 | - | 2 | 165 | |||
| AY358887.1 | - | 2 | 100 | |||
| BC027918.1 | - | 2 | 102 | 5226781 | ||
| BP331359.1 | - | 2 | 196 | |||
| CR450326.1 | - | 2 | 95 | |||
| U88320.1 | - | 2 | 109 | |||

CCL21
Genomic Map