siRNA Details
| Gene Symbol: C1orf162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300835.1 | NP_001287764.1 | 4 | 355 | ||
| NM_174896.3 | NP_777556.1 | 5 | 450 | |||
| XM_011540712.2 | XP_011539014.1 | 6 | 5930 | |||
| XM_017000324.1 | XP_016855813.1 | 5 | 5835 | |||
| GenBank mRNA | BC017973.1 | - | 5 | 337 | 4693393 | |
| HX785231.1 | - | 4 | 237 | |||

C1orf162
Genomic Map