siRNA Details
| Gene Symbol: LDLRAD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304263.1 | NP_001291192.1 | 5 | 1636 | ||
| NM_001304264.1 | NP_001291193.1 | 6 | 1752 | |||
| NM_174902.3 | NP_777562.1 | 6 | 1783 | |||
| GenBank mRNA | AK075546.1 | - | 6 | 1702 | ||
| BC042754.1 | - | 6 | 1686 | 4828259 | ||
| BC136286.1 | - | 6 | 1783 | 9020273 | ||
| BC143823.1 | - | 6 | 1752 | |||
| BC143824.1 | - | 5 | 1636 | 9052339 | ||

LDLRAD3
Genomic Map