siRNA Details
| Gene Symbol: PLEKHA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175058.4 | NP_778228.3 | 4, 5 | 329 | ||
| XM_017017240.1 | XP_016872729.1 | 4, 5 | 336 | |||
| XM_017017241.1 | XP_016872730.1 | 4, 5 | 336 | |||
| XM_017017242.1 | XP_016872731.1 | 4, 5 | 336 | |||
| XM_017017243.1 | XP_016872732.1 | 4, 5 | 336 | |||
| GenBank mRNA | BC071599.1 | - | 4, 5 | 325 | 30343461 | |
| CN293588.1 | - | 4, 5 | 329 | |||

PLEKHA7
Genomic Map