siRNA Details
| Gene Symbol: SYT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175733.3 | NP_783860.1 | 7 | 3175 | ||
| XM_011519901.2 | XP_011518203.1 | 8 | 3304 | |||
| GenBank mRNA | AK055003.1 | - | 6 | 2408 | ||
| AK055929.1 | - | 1 | 1182 | |||
| AK127795.1 | - | 1 | 2371 | |||
| AL137512.1 | - | 1 | 463 | |||
| BC046367.1 | - | 7 | 3175 | 5770087 | ||

SYT9
Genomic Map