siRNA Details
| Gene Symbol: DRAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178454.4 | NP_848549.3 | 9 | 1781 | ||
| XM_005270469.2 | XP_005270526.1 | 10 | 1839 | |||
| XM_005270470.1 | XP_005270527.1 | 10 | 1962 | |||
| XM_006710361.1 | XP_006710424.1 | 10 | 1799 | |||
| XM_006710362.1 | XP_006710425.1 | 10 | 1772 | |||
| XM_011540707.1 | XP_011539009.1 | 10 | 1780 | |||
| XM_011540708.1 | XP_011539010.1 | 10 | 1860 | |||
| XM_017000320.1 | XP_016855809.1 | 9 | 1723 | |||
| XM_017000321.1 | XP_016855810.1 | 9 | 1582 | |||
| GenBank mRNA | AK075350.1 | - | 5 | 1232 | ||
| AY336747.1 | - | 9 | 1660 | |||

DRAM2
Genomic Map