siRNA Details
| Gene Symbol: MFSD4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181644.4 | NP_857595.3 | 10 | 2231 | ||
| XM_011509229.2 | XP_011507531.1 | 9 | 1970 | |||
| GenBank mRNA | AK057733.1 | - | 6 | 1301 | ||
| AK090831.1 | - | 4 | 958 | |||
| AK091896.1 | - | 10 | 2172 | |||
| AK127321.1 | - | 2 | 1867 | |||
| AL713676.1 | - | 8 | 1537 | |||
| AY358107.1 | - | 8 | 1618 | |||
| BC036549.2 | - | 10 | 2179 | 5264261 | ||

MFSD4A
Genomic Map