siRNA Details
| Gene Symbol: DEUP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181645.3 | NP_857596.2 | 7 | 716 | ||
| XM_005273802.1 | XP_005273859.1 | 7 | 763 | |||
| XM_011542629.2 | XP_011540931.1 | 7 | 746 | |||
| XM_011542630.1 | XP_011540932.1 | 7 | 763 | |||
| XM_011542631.2 | XP_011540933.1 | 7 | 1823 | |||
| XM_011542632.2 | XP_011540934.1 | 7 | 746 | |||
| XM_011542633.2 | XP_011540935.1 | 7 | 746 | |||
| XM_011542635.2 | XP_011540937.1 | 7 | 746 | |||
| XM_011542637.2 | XP_011540939.1 | 7 | 746 | |||
| XM_017017245.1 | XP_016872734.1 | 7 | 746 | |||
| GenBank mRNA | AK058122.1 | - | 5 | 450 | ||
| AK127973.1 | - | 7 | 763 | |||
| BC017929.1 | - | 7 | 695 | |||

DEUP1
Genomic Map