siRNA Details
| Gene Symbol: CALHM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129742.1 | NP_001123214.1 | 3 | 1457 | ||
| GenBank mRNA | BC039499.1 | - | 2 | 851 | ||
| BC043367.1 | - | 3 | 1475 | 5110383 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CALHM3| Gene Symbol: CALHM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129742.1 | NP_001123214.1 | 3 | 1457 | ||
| GenBank mRNA | BC039499.1 | - | 2 | 851 | ||
| BC043367.1 | - | 3 | 1475 | 5110383 | ||