siRNA Details
Gene Symbol: KNCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097611.1 | NP_001091080.1 | 3 | 786 | ||
NM_001322255.1 | NP_001309184.1 | 4 | 1297 | |||
GenBank mRNA | AK056573.1 | - | 3 | 714 | ||
BC101295.1 | - | 3 | 708 | |||
BC101296.1 | - | 3 | 708 | |||
BC101297.2 | - | 3 | 708 | |||
BC101298.2 | - | 3 | 708 |
