siRNA Details
| Gene Symbol: FAAP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146310.1 | NP_001139782.1 | 8 | 2239 | ||
| NM_001256945.1 | NP_001243874.1 | 3 | 1369 | |||
| NM_001282671.1 | NP_001269600.1 | 8 | 2320 | |||
| NM_001282672.1 | NP_001269601.1 | 7 | 2189 | |||
| NM_001282673.1 | NP_001269602.1 | 4 | 1505 | |||
| XM_017000554.1 | XP_016856043.1 | 4 | 1480 | |||
| GenBank mRNA | AF469129.1 | - | 8 | 2286 | ||
| AK055593.1 | - | 7 | 2189 | |||
| AK127994.1 | - | 8 | 2239 | |||

FAAP20
Genomic Map