siRNA Details
| Gene Symbol: LINC01559 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036555.1 | 3 | 840 | |||
| GenBank mRNA | AK091129.1 | - | 3 | 806 | ||
| BC101220.1 | - | 3 | 657 | 40022214 | ||
| BC101222.2 | - | 3 | 657 | 40022216 | ||
| BC101223.1 | - | 3 | 657 | 40022217 | ||

LINC01559
Genomic Map