siRNA Details
| Gene Symbol: SECTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003004.2 | NP_002995.1 | 5 | 1327 | ||
| XM_005256392.3 | XP_005256449.1 | 5 | 1111 | |||
| XM_011523588.2 | XP_011521890.1 | 5 | 1358 | |||
| GenBank mRNA | AK223068.1 | - | 5 | 1020 | ||
| AK223104.1 | - | 5 | 1051 | |||
| BC017716.1 | - | 5 | 1021 | 4413387 | ||
| BI861812.1 | - | 3 | 748 | |||
| U77643.1 | - | 5 | 1047 | |||

SECTM1
Genomic Map