siRNA Details
| Gene Symbol: CYB561D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134400.1 | NP_001127872.1 | 3 | 1590 | ||
| NM_001134402.1 | NP_001127874.1 | 2 | 1396 | |||
| NM_001134403.1 | NP_001127875.1 | 4 | 1628 | |||
| NM_001134404.1 | NP_001127876.1 | 4 | 1705 | |||
| NM_182580.2 | NP_872386.1 | 3 | 1524 | |||
| XM_005270775.2 | XP_005270832.1 | 3 | 1601 | |||
| XM_005270776.3 | XP_005270833.1 | 3 | 1440 | |||
| XM_005270777.2 | XP_005270834.1 | 2 | 1480 | |||
| XM_011541286.2 | XP_011539588.1 | 4 | 1589 | |||
| XM_011541287.2 | XP_011539589.2 | 3 | 2717 | |||
| XM_017001078.1 | XP_016856567.1 | 4 | 1512 | |||
| XM_017001079.1 | XP_016856568.1 | 3 | 1334 | |||
| GenBank mRNA | AK096354.1 | - | 3 | 1503 | ||
| AK128798.1 | - | 4 | 1669 | |||
| BX647509.1 | - | 4 | 1608 | |||

CYB561D1
Genomic Map