siRNA Details
| Gene Symbol: C1orf111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182581.3 | NP_872387.2 | 3 | 841 | ||
| XM_005245103.3 | XP_005245160.1 | 2 | 835 | |||
| XM_011509443.2 | XP_011507745.1 | 3 | 1006 | |||
| XM_011509444.1 | XP_011507746.1 | 3 | 853 | |||
| GenBank mRNA | BC032957.1 | - | 3 | 848 | 4827863 | |

C1orf111
Genomic Map