siRNA Details
| Gene Symbol: FAM159A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135156.1 | 4, 5 | 753 | |||
| NR_135157.1 | 3, 4 | 660 | ||||
| GenBank mRNA | AK128236.1 | - | 3, 4 | 660 | ||
| BC041608.1 | - | 4, 5 | 703 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM159A| Gene Symbol: FAM159A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135156.1 | 4, 5 | 753 | |||
| NR_135157.1 | 3, 4 | 660 | ||||
| GenBank mRNA | AK128236.1 | - | 3, 4 | 660 | ||
| BC041608.1 | - | 4, 5 | 703 | |||