siRNA Details
| Gene Symbol: FAM159A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135156.1 | 6 | 978 | |||
| NR_135157.1 | 5 | 966 | ||||
| GenBank mRNA | AK128236.1 | - | 5 | 966 | ||
| BC041608.1 | - | 6 | 928 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM159A| Gene Symbol: FAM159A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135156.1 | 6 | 978 | |||
| NR_135157.1 | 5 | 966 | ||||
| GenBank mRNA | AK128236.1 | - | 5 | 966 | ||
| BC041608.1 | - | 6 | 928 | |||