siRNA Details
| Gene Symbol: SEMG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003007.4 | NP_002998.1 | 2 | 186 | ||
| GenBank mRNA | BC005229.1 | - | 2 | 117 | ||
| BC007096.1 | - | 2 | 117 | 4251523 | ||
| BC055416.1 | - | 2 | 117 | 6668871 | ||
| BP325874.1 | - | 2 | 186 | |||
| BT007177.1 | - | 2 | 96 | |||
| DA874748.1 | - | 2 | 153 | |||
| J04440.1 | - | 2 | 102 | |||

SEMG1
Genomic Map