siRNA Details
| Gene Symbol: SASS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304829.1 | NP_001291758.1 | 7 | 1020 | ||
| NM_194292.2 | NP_919268.1 | 8 | 1100 | |||
| XM_017000486.1 | XP_016855975.1 | 8 | 866 | |||
| GenBank mRNA | AK302513.1 | - | 9 | 850 | ||
| AK307824.1 | - | 7 | 887 | |||
| AL834265.1 | - | 4 | 363 | |||
| AY359522.1 | - | 8 | 960 | |||
| BC101026.3 | - | 7 | 813 | 40007432 | ||
| BX641109.1 | - | 8 | 919 | |||
| BX648406.1 | - | 8 | 856 | |||

SASS6
Genomic Map