siRNA Details
| Gene Symbol: ZBTB41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194314.2 | NP_919290.2 | 1 | 841 | ||
| NR_135153.1 | 1 | 841 | ||||
| XM_011509507.2 | XP_011507809.1 | 1 | 820 | |||
| XM_017001205.1 | XP_016856694.1 | 1 | 820 | |||
| GenBank mRNA | AK128549.1 | - | 1 | 820 | ||
| AY163816.2 | - | 1 | 841 | |||
| BC132818.1 | - | 1 | 805 | 40146840 | ||
| BC136600.1 | - | 1 | 805 | 9020590 | ||

ZBTB41
Genomic Map